2-Piperidinonicotinic acid ≥95%

Supplier: Matrix Scientific

045194-500MG 045194-1G 045194-5G
101869-622EA 181.54 USD
101869-622 101869-624 101869-626
2-Piperidinonicotinic acid ≥95%
2-Piperidinonicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MW: 206.25 g/mol
Melting Pt: 102…104 °C
MDL Number: MFCD02181988
CAS Number: 78253-61-9

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR