2-(Dimethoxymethyl)-5-hydroxy-1-benzofuran-3-carbaldehyde ≥95%
Supplier: Matrix Scientific
045180-500MG
101869-550EA
747.69
USD
101869-550
2-(Dimethoxymethyl)-5-hydroxy-1-benzofuran-3-carbaldehyde ≥95%
2-(Dimethoxymethyl)-5-hydroxy-1-benzofuran-3-carbaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₂O₅ Melting Pt: 137…139 °C |
MDL Number:
MFCD11553024 CAS Number: 1092352-23-2 |
Specification Test Results
Melting point | 137-139°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...