2-Chloro-1-(3,4-dibromo-2-thienyl)-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Chloroacetyl)-3,4-dibromothiophene
045176-1G
045176-500MG
045176-5G
101869-530EA
375.38
USD
101869-530
101869-532
101869-534
2-Chloro-1-(3,4-dibromo-2-thienyl)-1-ethanone ≥95%
2-Chloro-1-(3,4-dibromo-2-thienyl)-1-ethanone
Hazard: Irritant
Formula:
C₆H₃Br₂ClOS MW: 318.43 g/mol Melting Pt: 96…98 °C |
MDL Number:
MFCD06411631 CAS Number: 62673-71-6 |
Specification Test Results
Melting point | 96-98°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...