5-Methyl-3-[3-(trifluoromethyl)phenyl]-1H-pyrazole ≥95%

Supplier: Matrix Scientific

045173-500MG 045173-1G 045173-5G
101869-512EA 556.92 USD
101869-512 101869-514 101869-516
5-Methyl-3-[3-(trifluoromethyl)phenyl]-1H-pyrazole ≥95%
5-Methyl-3-[3-(trifluoromethyl)phenyl]-1H-pyrazole

Hazard: Irritant

Formula: C₁₁H₉F₃N₂
Melting Pt: 89…92 °C
MDL Number: MFCD06659570
CAS Number: 521267-21-0

Order Now

Specification Test Results

Melting point 89-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR