5-Nitrobenzoxazole ≥95%

Supplier: Matrix Scientific

045160-500MG 045160-1G 045160-5G
101869-440EA 532.31 USD
101869-440 101869-436 101869-438
5-Nitrobenzoxazole ≥95%
5-Nitrobenzoxazole

Hazard: Irritant

Formula: C₇H₄N₂O₃
MW: 164.1183 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD01359855
CAS Number: 70886-33-8

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR