2-(Chloromethyl)-5-nitrobenzoxazole ≥95%

Supplier: Matrix Scientific

045145-1G 045145-5G 045145-500MG
101869-354EA 375.38 USD
101869-354 101869-356 101869-358
2-(Chloromethyl)-5-nitrobenzoxazole ≥95%
2-(Chloromethyl)-5-nitrobenzoxazole

Hazard: Irritant

Formula: C₈H₅ClN₂O₃
Melting Pt: 115…117 °C
MDL Number: MFCD01596292
CAS Number: 119198-10-6

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR