Methyl-3-chloro-1H-indole-2-carboxylate ≥95%
Supplier: Matrix Scientific
045141-1G
045141-500MG
045141-5G
101869-330EA
192.31
USD
101869-330
101869-332
101869-334
Methyl-3-chloro-1H-indole-2-carboxylate ≥95%
Methyl-3-chloro-1H-indole-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₈ClNO₂ MW: 209.63 g/mol Melting Pt: 168…170 °C |
MDL Number:
MFCD04125889 CAS Number: 220664-32-4 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...