6-[(E)-(Dimethylamino)methylidene]-1,3-dimethylfuro[2,3-d]pyrimidine-2,4,5(1H,3H,6H)-trione ≥95%

Supplier: Matrix Scientific

045126-1G 045126-500MG 045126-5G
101869-262EA 181.54 USD
101869-262 101869-264 101869-266
6-[(E)-(Dimethylamino)methylidene]-1,3-dimethylfuro[2,3-d]pyrimidine-2,4,5(1H,3H,6H)-trione ≥95%
6-[(E)-(Dimethylamino)methylidene]-1,3-dimethylfuro[2,3-d]pyrimidine-2,4,5(1H,3H,6H)-trione

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₄
MW: 251.24 g/mol
Melting Pt: 248…250 °C
MDL Number: MFCD06200980
CAS Number: 453557-63-6

Order Now

Specification Test Results

Melting point 248-250°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR