Ethyl-1-(4-aminophenyl)-4-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
045120-1G
045120-5G
101869-240EA
401.54
USD
101869-240
101869-238
Ethyl-1-(4-aminophenyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(4-aminophenyl)-4-piperidinecarboxylate
Hazard: Irritant/Store in Freezer
Formula:
C₁₄H₂₀N₂O₂ Melting Pt: 40…42 °C |
MDL Number:
MFCD03425793 CAS Number: 439095-52-0 |
Specification Test Results
Melting point | 40-42°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...