4-(1-Benzothiophen-2-yl)-2-pyrimidinethiol ≥95%

Supplier: Matrix Scientific

045119-500MG
101869-234EA 1109.23 USD
101869-234
4-(1-Benzothiophen-2-yl)-2-pyrimidinethiol ≥95%
4-(1-Benzothiophen-2-yl)-2-pyrimidinethiol

Hazard: Irritant

Formula: C₁₂H₈N₂S₂
Melting Pt: 272…274 °C
MDL Number: MFCD03787202
CAS Number: 866049-73-2

Order Now

Specification Test Results

Melting point 272-274°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR