1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-pyrrole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
045118-1G
045118-500MG
045118-5G
101869-230EA
467.69
USD
101869-230
101869-232
101869-228
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-pyrrole-2-carbaldehyde ≥95%
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₉H₇ClN₂OS Melting Pt: 58…60 °C |
MDL Number:
MFCD03102582 CAS Number: 692287-27-7 |
Specification Test Results
Melting point | 58-60°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...