1-(4-Acetylphenyl)-1H-pyrrole-2,5-dione ≥95%

Supplier: Matrix Scientific

045115-500MG 045115-5G 045115-1G
101869-212EA 278.46 USD
101869-212 101869-214 101869-216
1-(4-Acetylphenyl)-1H-pyrrole-2,5-dione ≥95%
1-(4-Acetylphenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₂H₉NO₃
Melting Pt: 151…153 °C
MDL Number: MFCD00014537
CAS Number: 1082-85-5

Order Now

Specification Test Results

Melting point 151-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR