4-Bromo-5-isopropyl-2-methylbenzenol ≥95%

Supplier: Matrix Scientific

045108-5G 045108-500MG 045108-1G
101869-188EA 238.46 USD
101869-188 101869-190 101869-192
4-Bromo-5-isopropyl-2-methylbenzenol ≥95%
4-Bromo-5-isopropyl-2-methylbenzenol

Hazard: Irritant

Formula: C₁₀H₁₃BrO
Melting Pt: 28…31 °C
MDL Number: MFCD03791230
CAS Number: 121665-99-4

Order Now

Specification Test Results

Melting point 28-31°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR