3-[4-(Benzyloxy)phenyl]-2-phenylacrylic acid ≥95%
Supplier: Matrix Scientific
045096-1G
045096-500MG
045096-5G
101869-156EA
278.46
USD
101869-156
101869-158
101869-160
3-[4-(Benzyloxy)phenyl]-2-phenylacrylic acid ≥95%
3-[4-(Benzyloxy)phenyl]-2-phenylacrylic acid
Hazard: Irritant
Formula:
C₂₂H₁₈O₃ Melting Pt: 218…221 °C |
MDL Number:
MFCD03001274 CAS Number: 126582-18-1 |
Specification Test Results
Melting point | 218-221°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...