2-(2-Methoxyphenyl)-3-phenylacrylic acid ≥95%
Supplier: Matrix Scientific
045094-5G
045094-1G
045094-500MG
101869-150EA
278.46
USD
101869-150
101869-152
101869-154
2-(2-Methoxyphenyl)-3-phenylacrylic acid ≥95%
2-(2-Methoxyphenyl)-3-phenylacrylic acid
Hazard: Irritant
Formula:
C₁₆H₁₄O₃ Melting Pt: 146…148 °C |
MDL Number:
MFCD03001273 CAS Number: 1657-65-4 |
Specification Test Results
Melting point | 146-148°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...