2,4-Dihydroxy-3,3-dimethylbutanamide ≥95%

Supplier: Matrix Scientific

045093-500MG 045093-1G 045093-5G
101869-144EA 278.46 USD
101869-144 101869-146 101869-148
2,4-Dihydroxy-3,3-dimethylbutanamide ≥95%
2,4-Dihydroxy-3,3-dimethylbutanamide

Hazard: Irritant

Formula: C₆H₁₃NO₃
Melting Pt: 126…128 °C
MDL Number: MFCD00825730
CAS Number: 4417-86-1

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR