Methyl-3-(2-methoxyphenyl)-2-phenylacrylate ≥95%
Supplier: Matrix Scientific
045091-5G
045091-1G
045091-500MG
101869-132EA
375.38
USD
101869-132
101869-134
101869-136
Methyl-3-(2-methoxyphenyl)-2-phenylacrylate ≥95%
Methyl-3-(2-methoxyphenyl)-2-phenylacrylate
Hazard: Irritant
Formula:
C₁₇H₁₆O₃ Melting Pt: 102…103 °C |
MDL Number:
MFCD03001272 CAS Number: 42307-45-9 |
Specification Test Results
Melting point | 102-103°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...