1-(6-Methyl-2-pyridinyl)-1H-imidazole-4-carbohydrazide ≥95%

Supplier: Matrix Scientific

045085-5G 045085-500MG 045085-1G
101869-096EA 747.69 USD
101869-096 101869-098 101869-100
1-(6-Methyl-2-pyridinyl)-1H-imidazole-4-carbohydrazide ≥95%
1-(6-Methyl-2-pyridinyl)-1H-imidazole-4-carbohydrazide

Hazard: Irritant

Formula: C₁₀H₁₁N₅O
Melting Pt: 182…184 °C
MDL Number: MFCD02083045
CAS Number: 478063-72-8

Order Now

Specification Test Results

Melting point 182-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR