N-(1H-Indazol-6-yl)thiourea ≥95%

Supplier: Matrix Scientific

045080-1G 045080-500MG 045080-5G
101869-074EA 278.46 USD
101869-074 101869-076 101869-078
N-(1H-Indazol-6-yl)thiourea ≥95%
N-(1H-Indazol-6-yl)thiourea

Hazard: Irritant

Formula: C₈H₈N₄S
Melting Pt: 207 °C
MDL Number: MFCD03102726
CAS Number: 99055-55-7

Order Now

Specification Test Results

Melting point 207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR