1-(4-Chlorophenyl)-3-[imino(nitroamino)methyl]thiourea ≥95%

Supplier: Matrix Scientific

045079-500MG 045079-1G 045079-5G
101869-068EA 181.54 USD
101869-068 101869-070 101869-072
1-(4-Chlorophenyl)-3-[imino(nitroamino)methyl]thiourea ≥95%
1-(4-Chlorophenyl)-3-[imino(nitroamino)methyl]thiourea

Hazard: Irritant

Formula: C₈H₈ClN₅O₂S
Melting Pt: 158…160 °C
MDL Number: MFCD08056646
CAS Number: 204188-36-3

Order Now

Specification Test Results

Melting point 158-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR