2-(2,6-Difluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%

Supplier: Matrix Scientific

045064-500MG
101868-988EA 556.92 USD
101868-988
2-(2,6-Difluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%
2-(2,6-Difluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile

Hazard: Irritant

Formula: C₁₀H₄F₂N₄O₂
Melting Pt: 235…238 °C
MDL Number: MFCD00794280
CAS Number: 338982-41-5

Order Now

Specification Test Results

Melting point 235-238°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR