2-[Cyano(2,4-dichlorophenyl)methyl]-6-fluorobenzenecarbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
2-[Cyano(2,4-dichlorophenyl)methyl]-6-fluorobenzonitrile
045052-5G
045052-1G
045052-500MG
101868-918EA
181.54
USD
101868-918
101868-920
101868-922
2-[Cyano(2,4-dichlorophenyl)methyl]-6-fluorobenzenecarbonitrile ≥95%
2-[Cyano(2,4-dichlorophenyl)methyl]-6-fluorobenzenecarbonitrile
Hazard: Irritant
Formula:
C₁₅H₇Cl₂FN₂ MW: 305.14 g/mol Melting Pt: 102…104 °C |
MDL Number:
MFCD00231988 CAS Number: 338965-45-0 |
Specification Test Results
Melting point | 102-104°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...