(4-Nitrobenzyl)malononitrile ≥95%

Supplier: Matrix Scientific

045051-1G 045051-500MG 045051-5G
101868-916EA 802.7 USD
101868-916 101868-914 101868-912
(4-Nitrobenzyl)malononitrile ≥95%
(4-Nitrobenzyl)malononitrile

Hazard: Irritant

Formula: C₁₀H₇N₃O₂
Melting Pt: 153…155 °C
MDL Number: MFCD00231640
CAS Number: 6731-53-9

Order Now

Specification Test Results

Melting point 153-155°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR