3-(Benzyloxy)-2-methyl-4(1H)-pyridinone ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Benzyloxy)-2-methylpyridin-4(1H)-one
045049-5G
045049-1G
101868-906EA
96.92
USD
101868-906
101868-908
3-(Benzyloxy)-2-methyl-4(1H)-pyridinone ≥95%
3-(Benzyloxy)-2-methyl-4(1H)-pyridinone
Hazard: Irritant
Formula:
C₁₃H₁₃NO₂ MW: 215.25 g/mol Melting Pt: 171…173 °C |
MDL Number:
MFCD00215656 CAS Number: 61160-18-7 |
Specification Test Results
Melting point | 171-173°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...