6-Amino-5-(4-methoxybenzyl)-2-phenyl-4-pyrimidinylamine ≥95%

Supplier: Matrix Scientific

045045-1G 045045-500MG 045045-5G
101868-880EA 747.69 USD
101868-880 101868-884 101868-882
6-Amino-5-(4-methoxybenzyl)-2-phenyl-4-pyrimidinylamine ≥95%
6-Amino-5-(4-methoxybenzyl)-2-phenyl-4-pyrimidinylamine

Hazard: Irritant

Formula: C₁₈H₁₈N₄O
Melting Pt: 181…183 °C
MDL Number: MFCD00231625
CAS Number: 163590-00-9

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR