Ethyl-8-hydroxy[1,3]dioxolo[4,5-g]quinoline-7-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 8-hydroxy-[1,3]dioxolo[4,5-g]quinoline-7-carboxylate
045035-5G
045035-1G
045035-500MG
101868-822EA
375.38
USD
101868-822
101868-826
101868-824
Ethyl-8-hydroxy[1,3]dioxolo[4,5-g]quinoline-7-carboxylate ≥95%
Ethyl-8-hydroxy[1,3]dioxolo[4,5-g]quinoline-7-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₁NO₅ MW: 261.24 g/mol Melting Pt: 311…313 °C (dec.) |
MDL Number:
MFCD00173400 CAS Number: 14205-65-3 |
Specification Test Results
Melting point | 311-313°C(d |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...