Ethyl-2-(2-bromoacetyl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(2-bromoacetyl)benzoate
045006-1G
045006-500MG
045006-5G
101868-682EA
493.85
USD
101868-682
101868-680
101868-684
Ethyl-2-(2-bromoacetyl)benzenecarboxylate ≥95%
Ethyl-2-(2-bromoacetyl)benzenecarboxylate
Hazard: Irritant/Store in Freezer
Formula:
C₁₁H₁₁BrO₃ MW: 271.12 g/mol Melting Pt: 60…62 °C |
MDL Number:
MFCD00142009 CAS Number: 133993-34-7 |
Specification Test Results
Melting point | 60-62°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...