Methyl-2-(3-ethoxy-3-oxopropanimidoyl)-1-hydrazinecarboxylate ≥95%
Supplier: Matrix Scientific
045004-1G
045004-5G
045004-500MG
101868-670EA
747.69
USD
101868-670
101868-674
101868-672
Methyl-2-(3-ethoxy-3-oxopropanimidoyl)-1-hydrazinecarboxylate ≥95%
Methyl-2-(3-ethoxy-3-oxopropanimidoyl)-1-hydrazinecarboxylate
Hazard: Irritant
Formula:
C₇H₁₃N₃O₄ Melting Pt: 120…122 °C |
MDL Number:
MFCD00664782 CAS Number: 338791-53-0 |
Specification Test Results
Melting point | 120-122°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...