5-(4-Chlorophenyl)furfural ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(4-Chlorophenyl)-2-furaldehyde5-(4-Chlorophenyl)furan-2-carbaldehyde

044994-1G
101868-628EA 407.69 USD
101868-628
5-(4-Chlorophenyl)furfural ≥95%
5-(4-Chlorophenyl)furfural

Hazard: Irritant

Formula: C₁₁H₇ClO₂
MW: 206.625 g/mol
Melting Pt: 132…134 °C
MDL Number: MFCD00195947
CAS Number: 34035-03-5

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR