6-Oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
1-[4-(Trifluoromethyl)benzyl]pyridin-2-one-5-carboxylic acid
044992-1G
044992-5G
044992-500MG
101868-620EA
207.69
USD
101868-620
101868-624
101868-622
6-Oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid ≥97%
6-Oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₀F₃NO₃ MW: 297.24 g/mol Melting Pt: 251…253 °C |
MDL Number:
MFCD00243721 CAS Number: 338783-75-8 |
Specification Test Results
Melting point | 251-253°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...