2-Oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[4-(Trifluoromethyl)benzyl]pyridin-2-one-3-carboxylic acid

044988-1G 044988-5G 044988-500MG
101868-598EA 698.46 USD
101868-598 101868-596 101868-600
2-Oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%
2-Oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₀F₃NO₃
MW: 297.24 g/mol
Melting Pt: 192…193 °C
MDL Number: MFCD00243696
CAS Number: 66158-46-1

Order Now

Specification Test Results

Melting point 192-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR