4-(2-Pyrazinyl)-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

044972-500MG 044972-1G 044972-5G
101868-514EA 747.69 USD
101868-514 101868-518 101868-516
4-(2-Pyrazinyl)-2-pyrimidinamine ≥95%
4-(2-Pyrazinyl)-2-pyrimidinamine

Hazard: Irritant

Formula: C₈H₇N₅
Melting Pt: 207…209 °C
MDL Number: MFCD04036165
CAS Number: 874792-15-1

Order Now

Specification Test Results

Melting point 207-209°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR