3-(Dimethylamino)-1-(1,3-thiazol-2-yl)-2-buten-1-one ≥95%
Supplier: Matrix Scientific
044969-5G
044969-500MG
044969-1G
101868-498EA
927.69
USD
101868-498
101868-496
101868-500
3-(Dimethylamino)-1-(1,3-thiazol-2-yl)-2-buten-1-one ≥95%
3-(Dimethylamino)-1-(1,3-thiazol-2-yl)-2-buten-1-one
Hazard: Irritant
Formula:
C₉H₁₂N₂OS MW: 196.27 g/mol Melting Pt: 110…112 °C |
MDL Number:
MFCD08056641 CAS Number: 886361-88-2 |
Specification Test Results
Melting point | 110-112°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...