7-Cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-7-cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidine

044949-5G 044949-500MG 044949-1G
101868-388EA 350.77 USD
101868-388 101868-386 101868-390
7-Cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
7-Cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Hazard: Irritant

Formula: C₈H₉N₅
MW: 175.19 g/mol
Melting Pt: 235…237 °C
MDL Number: MFCD07366548
CAS Number: 885949-41-7

Order Now

Specification Test Results

Melting point 235-237°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR