2-(3-Oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)acetic acid ≥95%

Supplier: Matrix Scientific

044941-500MG 044941-1G 044941-5G
101868-340EA 747.69 USD
101868-340 101868-344 101868-342
2-(3-Oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)acetic acid ≥95%
2-(3-Oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)acetic acid

Hazard: Irritant

Formula: C₁₀H₉NO₄
Melting Pt: 281…283 °C
MDL Number: MFCD06659563
CAS Number: 134997-91-4

Order Now

Specification Test Results

Melting point 281-283°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR