Methyl-4-(5-amino-2-pyridinyl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
044935-1G
044935-500MG
044935-5G
101868-304EA
747.69
USD
101868-304
101868-308
101868-306
Methyl-4-(5-amino-2-pyridinyl)benzenecarboxylate ≥95%
Methyl-4-(5-amino-2-pyridinyl)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₂ MW: 228.25 g/mol Melting Pt: 189…192 °C |
MDL Number:
MFCD06411623 CAS Number: 223127-54-6 |
Specification Test Results
Melting point | 189-192°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...