4-Acetyl-8-benzyl-1-thia-4,8-diazaspiro[4.5]decane-3-carboxylic acid hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Acetyl-8-benzyl-4,8-diaza-1-thiaspiro[4.5]decane-3-carboxylic acid hydrochloride
044918-5G
044918-1G
044918-500MG
101868-216EA
350.77
USD
101868-216
101868-214
101868-218
4-Acetyl-8-benzyl-1-thia-4,8-diazaspiro[4.5]decane-3-carboxylic acid hydrochloride ≥95%
4-Acetyl-8-benzyl-1-thia-4,8-diazaspiro[4.5]decane-3-carboxylic acid hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₃ClN₂O₃S MW: 370.9 g/mol Melting Pt: 251…253 °C |
MDL Number:
MFCD06795478 CAS Number: 1030253-99-6 |
Specification Test Results
Melting point | 251-253°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...