1-Boc-4-(hydroxymethyl)piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: N-BOC-4-Piperidinemethanoltert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate1-Boc-4-piperidinemethanol

044910-25G
101868-170EA 30.77 USD
101868-170
1-Boc-4-(hydroxymethyl)piperidine ≥95%
1-Boc-4-(hydroxymethyl)piperidine

Hazard: Irritant

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
Melting Pt: 78…82 °C
MDL Number: MFCD02094488
CAS Number: 123855-51-6

Order Now

Specification Test Results

Melting point 69-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR