4-[2-(4-Nitrophenoxy)ethyl]morpholine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-(4-Nitrophenoxy)ethyl)morpholine

044897-5G 044897-500MG 044897-1G
101868-110EA 181.54 USD
101868-110 101868-114 101868-112
4-[2-(4-Nitrophenoxy)ethyl]morpholine ≥95%
4-[2-(4-Nitrophenoxy)ethyl]morpholine

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
Melting Pt: 80…82 °C
MDL Number: MFCD00434241
CAS Number: 65300-53-0

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR