2-Chloro-5-isopropyl-1,3-thiazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
044894-500MG
044894-1G
044894-5G
101868-098EA
375.38
USD
101868-098
101868-102
101868-100
2-Chloro-5-isopropyl-1,3-thiazole-4-carboxylic acid ≥95%
2-Chloro-5-isopropyl-1,3-thiazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₇H₈ClNO₂S Melting Pt: 209…212 °C |
MDL Number:
MFCD03791228 CAS Number: 886360-70-9 |
Specification Test Results
Melting point | 209-212°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...