Dimethyl-2-[2-nitro-5-(trifluoromethyl)phenyl]malonate ≥95%

Supplier: Matrix Scientific

044888-500MG 044888-1G 044888-5G
101868-066EA 232.31 USD
101868-066 101868-064 101868-068
Dimethyl-2-[2-nitro-5-(trifluoromethyl)phenyl]malonate ≥95%
Dimethyl-2-[2-nitro-5-(trifluoromethyl)phenyl]malonate

Hazard: Irritant

Formula: C₁₂H₁₀F₃NO₆
Melting Pt: 64…65 °C
MDL Number: MFCD03787568
CAS Number: 866039-51-2

Order Now

Specification Test Results

Melting point 64-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR