3-(Trifluoroacetyl)indole ≥95%

Supplier: Matrix Scientific

044866-5G 044866-500MG 044866-1G
101867-960EA 532.31 USD
101867-960 101867-956 101867-958
3-(Trifluoroacetyl)indole ≥95%
3-(Trifluoroacetyl)indole

Hazard: Irritant

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
Melting Pt: 218…220 °C
MDL Number: MFCD00182114
CAS Number: 14618-45-2

Order Now

Specification Test Results

Melting point 218-220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR