Quinoxalin-6-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Aminoquinoxaline

044858-5G 044858-1G
101867-928EA 120 USD
101867-928 101867-926
Quinoxalin-6-amine ≥95%
Quinoxalin-6-amine

Hazard: Irritant

Formula: C₈H₇N₃
MW: 145.1600 g/mol
Melting Pt: 157…159 °C
MDL Number: MFCD00462821
CAS Number: 6298-37-9

Order Now

Specification Test Results

Melting point 157-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR