6-Ethyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone ≥95%

Supplier: Matrix Scientific

044853-1G 044853-500MG 044853-5G
101867-896EA 200 USD
101867-896 101867-898 101867-900
6-Ethyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone ≥95%
6-Ethyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone

Hazard: Irritant

Formula: C₆H₈N₂OS
Melting Pt: 228…230 °C
MDL Number: MFCD02186898
CAS Number: 53939-83-6

Order Now

Specification Test Results

Melting point 228-230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR