3-(2-Hydroxy-1-phenylethyl)-2-thioxotetrahydro-4H-imidazol-4-one ≥95%
Supplier: Matrix Scientific
044840-5G
044840-1G
044840-500MG
101867-830EA
927.69
USD
101867-830
101867-828
101867-832
3-(2-Hydroxy-1-phenylethyl)-2-thioxotetrahydro-4H-imidazol-4-one ≥95%
3-(2-Hydroxy-1-phenylethyl)-2-thioxotetrahydro-4H-imidazol-4-one
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O₂S Melting Pt: 153…155 °C |
MDL Number:
MFCD01316095 CAS Number: 338777-60-9 |
Specification Test Results
Melting point | 153-155°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...