2-[(3,5-Dichloro-2-pyridinyl)sulfonyl]acetic acid ≥95%
Supplier: Matrix Scientific
044829-5G
044829-500MG
044829-1G
101867-794EA
1600
USD
101867-794
101867-790
101867-792
2-[(3,5-Dichloro-2-pyridinyl)sulfonyl]acetic acid ≥95%
2-[(3,5-Dichloro-2-pyridinyl)sulfonyl]acetic acid
Hazard: Irritant
Formula:
C₇H₅Cl₂NO₄S Melting Pt: 150…154 °C |
MDL Number:
MFCD01569019 CAS Number: 338962-58-6 |
Specification Test Results
Melting point | 150-154°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...