4-(2,4-Difluorophenoxy)-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

044821-1G 044821-5G 044821-500MG
101867-750EA 350.77 USD
101867-750 101867-752 101867-748
4-(2,4-Difluorophenoxy)-3-nitrobenzenecarbaldehyde ≥95%
4-(2,4-Difluorophenoxy)-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₇F₂NO₄
Melting Pt: 88…90 °C
MDL Number: MFCD00794806
CAS Number: 173282-22-9

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR