Ethyl-4-(2-chloro-6-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(2-chloro-6-fluorophenyl)-6-methyl-1,2,3,4-tetrahydropyrid-2-one-5-carboxylate

044810-1G 044810-5G 044810-500MG
101867-696EA 806.15 USD
101867-696 101867-692 101867-694
Ethyl-4-(2-chloro-6-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%
Ethyl-4-(2-chloro-6-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₅ClFNO₃
MW: 311.74 g/mol
Melting Pt: 200…202 °C
MDL Number: MFCD00664286
CAS Number: 299920-90-4

Order Now

Specification Test Results

Melting point 200-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR