3,5-Dioxo-2-[3-(trifluoromethyl)phenyl]-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%
Supplier: Matrix Scientific
044807-1G
044807-5G
044807-500MG
101867-674EA
556.92
USD
101867-674
101867-676
101867-678
3,5-Dioxo-2-[3-(trifluoromethyl)phenyl]-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%
3,5-Dioxo-2-[3-(trifluoromethyl)phenyl]-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile
Hazard: Irritant
Formula:
C₁₁H₅F₃N₄O₂ MW: 282.18 g/mol Melting Pt: 173…175 °C |
MDL Number:
MFCD00274940 CAS Number: 121065-97-2 |
Specification Test Results
Melting point | 173-175°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...