4-[(5-Chloro-1,2,3-thiadiazol-4-yl)methoxy]benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
044788-500MG
044788-1G
101867-598EA
375.38
USD
101867-598
101867-600
4-[(5-Chloro-1,2,3-thiadiazol-4-yl)methoxy]benzenecarbaldehyde ≥95%
4-[(5-Chloro-1,2,3-thiadiazol-4-yl)methoxy]benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₀H₇ClN₂O₂S Melting Pt: 79…81 °C |
MDL Number:
MFCD00232349 CAS Number: 338954-22-6 |
Specification Test Results
Melting point | 79-81°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...